accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COCH3OOCH3(E)

CAS RN 15001-27-1

IUPAC Name: (3E)-4-(3,4-dimethoxyphenyl)but-3-en-2-one
Synonyms: (3E)-4-(3,4-dimethoxyphenyl)but-3-en-2-one
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: XUYBNDHXZMIALN-SNAWJCMRSA-N
SMILES: COC1=CC=C(\C=C\C(C)=O)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results