accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14997-58-1

OHONHNNHOO

CAS RN 14997-58-1

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-(3H-imidazol-4-yl)propanoic acid
Molecular Weight (g/mol): 289.29
Molecular Formula: C14H15N3O4
InChi Key: WCOJOHPAKJFUDF-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CN=CN1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)