accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClPO

CAS RN 1499-21-4

IUPAC Name: diphenylphosphinoyl chloride
Synonyms: diphenylphosphinoyl chloride
Molecular Weight (g/mol): 236.63
Molecular Formula: C12H10ClOP
InChi Key: QPQGTZMAQRXCJW-UHFFFAOYSA-N
SMILES: ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)