accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SSOOHCl

CAS RN 149609-86-9

IUPAC Name: 4-[(4-chlorophenyl)sulfanyl]thiophene-3-carboxylic acid
Synonyms: 4-[(4-chlorophenyl)sulfanyl]thiophene-3-carboxylic acid
Molecular Weight (g/mol): 270.75
Molecular Formula: C11H7ClO2S2
InChi Key: UKDSGGJQRSNTHC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC=C1SC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results