accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 149506-93-4

OHON

CAS RN 149506-93-4

IUPAC Name: 4'-cyano-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 4'-cyano-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: WLUGFHVTVXLVFB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results