accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 149506-25-2

CH3OOBr

CAS RN 149506-25-2

IUPAC Name: methyl 4'-bromo-[1,1'-biphenyl]-3-carboxylate
Synonyms: methyl 4'-bromo-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol): 291.14
Molecular Formula: C14H11BrO2
InChi Key: UTOKFKAMZICCBU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results