accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNFFF

CAS RN 14925-10-1

IUPAC Name: N-methyl-2-(trifluoromethyl)aniline
Synonyms: N-methyl-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: RNRDHZGWBBEZBK-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results