accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14916-79-1

CH3HO

CAS RN 14916-79-1

IUPAC Name: hept-3-yn-1-ol
Synonyms: hept-3-yn-1-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: PSWHODJVUOXHKA-UHFFFAOYSA-N
SMILES: CCCC#CCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)