accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14915-37-8

CuONSONS

CAS RN 14915-37-8

IUPAC Name: copper(2+) bis((1-oxidopyridin-1-ium-2-yl)sulfanide)
Synonyms: copper(2+) bis((1-oxidopyridin-1-ium-2-yl)sulfanide)
Molecular Weight (g/mol): 315.85
Molecular Formula: C10H8CuN2O2S2
InChi Key: FNQNNKLJHNPTGT-UHFFFAOYSA-L
SMILES: [Cu++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)