accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14913-20-3

OPOOOEu

CAS RN 14913-20-3

IUPAC Name: europium(3+) hydrate phosphate
Synonyms: europium(3+) hydrate phosphate
Molecular Weight (g/mol): 264.95
Molecular Formula: EuH2O5P
InChi Key: BOCFVANIXFYKBV-UHFFFAOYSA-K
SMILES: O.[Eu+3].[O-]P([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results