accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14913-14-5

OPOOOLa

CAS RN 14913-14-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 236.9
Molecular Formula: H3LaO4P
InChi Key: AUOXQXPFOXZHRO-UHFFFAOYSA-N
SMILES: [La].OP(O)(O)=O
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results