accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 148980-31-8

FFFSOOOFFFSOOOFFFSOOOTb

CAS RN 148980-31-8

IUPAC Name: terbium(3+) tritrifluoromethanesulfonate
Synonyms: terbium(3+) tritriflate
Molecular Weight (g/mol): 606.11
Molecular Formula: C3F9O9S3Tb
InChi Key: JLYAVYSVLLIWEK-UHFFFAOYSA-K
SMILES: [Tb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results