accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14892-14-9

HOOOFFFFF

CAS RN 14892-14-9

IUPAC Name: 2-(2,3,4,5,6-pentafluorophenoxy)acetic acid
Synonyms: pentafluorophenoxyacetic acid
Molecular Weight (g/mol): 242.1
Molecular Formula: C8H3F5O3
InChi Key: SMXPFEBIAASLOR-UHFFFAOYSA-N
SMILES: OC(=O)COC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results