accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrH3CP

CAS RN 14866-43-4

IUPAC Name: hexadecyltriphenylphosphanium bromide
Synonyms: hexadecyltriphenylphosphanium bromide
Molecular Weight (g/mol): 567.64
Molecular Formula: C34H48BrP
InChi Key: UXMZNEHSMYESLH-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results