accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOO

CAS RN 1486-51-7

IUPAC Name: 4-(benzyloxy)benzoic acid
Synonyms: 4-(benzyloxy)benzoic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: AQSCHALQLXXKKC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)