CAS RN 148516-68-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Amino-N-(1-ethyl-3-piperidinyl)-5-(ethylsulfonyl)-2-methoxybenzamide, TRC
CAS: 148516-68-1 Molecular Formula: C17 H27 N3 O4 S Molecular Weight (g/mol): 369.479 Synonym: Benzamide, 4-amino-N-(1-ethyl-3-piperidinyl)-5-(ethylsulfonyl)-2-methoxy-,4-Amino-N-(1-ethyl-3-piperidyl)-5-ethylsulfonyl-2-methoxy-benzamide,Amisulpride Imp G (EP),Amisulpride EP Impurity G IUPAC Name: 4-amino-N-(1-ethylpiperidin-3-yl)-5-ethylsulfonyl-2-methoxybenzamide SMILES: CCN1CCCC(C1)NC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC