accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14794-99-1

H3COCH3OSn

CAS RN 14794-99-1

IUPAC Name: λ²-tin(2+) dimethanolate
Synonyms: λ²-tin(2+) bis(methoxide)
Molecular Weight (g/mol): 180.78
Molecular Formula: C2H6O2Sn
InChi Key: UISUQHKSYTZXSF-UHFFFAOYSA-N
SMILES: [Sn++].C[O-].C[O-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)