accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 147834-57-9

CH3H3CH3COCH3CH3

CAS RN 147834-57-9

IUPAC Name: 1-(2,3,4,5,6-pentamethylphenyl)-2-phenylethan-1-one
Synonyms: 1-(2,3,4,5,6-pentamethylphenyl)-2-phenylethanone
Molecular Weight (g/mol): 266.38
Molecular Formula: C19H22O
InChi Key: CFSAHKOFVAHZDH-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C(=O)CC2=CC=CC=C2)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results