accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OBrO

CAS RN 147644-11-9

IUPAC Name: 7-bromo-3,4-dihydro-2H-1,5-benzodioxepine
Synonyms: 7-bromo-3,4-dihydro-2H-1,5-benzodioxepine
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: AZCHNKNSOZJHSH-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCCCO2)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)