CAS RN 147404-69-1
CAS RN 147404-69-1
IUPAC Name:
4'-methyl-[1,1'-biphenyl]-3-carboxylate
Synonyms:
4'-methyl-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol):
211.24
Molecular Formula:
C14H11O2
InChi Key:
APYJGQZXCGXMGB-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C=C1)C1=CC=CC(=C1)C([O-])=O
1
–
1
of
1
results
1
4'-Methylbiphenyl-3-carboxylic acid, 95%, Thermo Scientific™
CAS: 147404-69-1 Molecular Formula: C14H11O2 Molecular Weight (g/mol): 211.24 MDL Number: MFCD00452725 InChI Key: APYJGQZXCGXMGB-UHFFFAOYSA-M Synonym: 4'-methylbiphenyl-3-carboxylic acid,4'-methyl-1,1'-biphenyl-3-carboxylic acid,3-4-methylphenyl benzoic acid,4'-methyl-biphenyl-3-carboxylic acid,4'-methyl-3-biphenylcarboxylic acid,4'-methyl 1,1'-biphenyl-3-carboxylic acid,1,1'-biphenyl-3-carboxylicacid, 4'-methyl,pubchem9064,acmc-20aih3,amtda023 PubChem CID: 2773978 IUPAC Name: 3-(4-methylphenyl)benzoic acid SMILES: CC1=CC=C(C=C1)C1=CC=CC(=C1)C([O-])=O