accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14731-10-3

IUPAC Name: 3-(chloromethyl)-5-phenyl-1,2-oxazole
Synonyms: 3-(chloromethyl)-5-phenyl-1,2-oxazole
Molecular Weight (g/mol): 193.63
Molecular Formula: C10H8ClNO
InChi Key: MLJJRVMANUGETQ-UHFFFAOYSA-N
SMILES: ClCC1=NOC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results