accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 147290-11-7

H2COOHNOHONHOO

CAS RN 147290-11-7

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 438.48
Molecular Formula: C24H26N2O6
InChi Key: RXLIOYNXBHZZBI-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)