accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOSNH2

CAS RN 147031-85-4

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-2-carbothioamide
Synonyms: 2,3-dihydro-1,4-benzodioxine-2-carbothioamide
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: ZOHVHNZPAGHKKO-UHFFFAOYNA-N
SMILES: NC(=S)C1COC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results