accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 146941-72-2

IUPAC Name: 4-bromo-1-methyl-1H-pyrazol-3-amine
Synonyms: 4-bromo-1-methylpyrazol-3-amine
Molecular Weight (g/mol): 176.02
Molecular Formula: C4H6BrN3
InChi Key: WYPUMGAZSVZUGJ-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(N)=N1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)