accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14678-02-5

IUPAC Name: 3-methyl-1,2-oxazol-5-amine
Synonyms: 3-methyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: FNXYWHTZDAVRTB-UHFFFAOYSA-N
SMILES: CC1=NOC(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)