accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 146548-59-6

H3COOH3CNH2OCH3HCl

CAS RN 146548-59-6

IUPAC Name: 1-(2,4,6-trimethoxyphenyl)methanamine hydrochloride
Synonyms: 1-(2,4,6-trimethoxyphenyl)methanamine hydrochloride
Molecular Weight (g/mol): 233.69
Molecular Formula: C10H16ClNO3
InChi Key: BLFRMOOGAICNSZ-UHFFFAOYSA-N
SMILES: Cl.COC1=CC(OC)=C(CN)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results