accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14651-42-4

Br

CAS RN 14651-42-4

IUPAC Name: 1-(bromomethyl)adamantane
Synonyms: 1-(bromomethyl)adamantane
Molecular Weight (g/mol): 229.16
Molecular Formula: C11H17Br
InChi Key: RLRBYCAVZBQANU-UHFFFAOYSA-N
SMILES: BrCC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results