accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 146449-90-3

BOCH3OHHO

CAS RN 146449-90-3

IUPAC Name: [4-(pentyloxy)phenyl]boronic acid
Synonyms: 4-(pentyloxy)phenylboronic acid
Molecular Weight (g/mol): 208.06
Molecular Formula: C11H17BO3
InChi Key: WHFKSJLAGACWMF-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results