accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14637-88-8

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHDy

CAS RN 14637-88-8

IUPAC Name: tris(4-hydroxypent-3-en-2-one) dysprosium
Synonyms: dysprosium acetylacetonate
Molecular Weight (g/mol): 462.85
Molecular Formula: C15H24DyO6
InChi Key: BYBFJHANLYDGFD-UHFFFAOYSA-N
SMILES: [Dy].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results