accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 146195-69-9

H3COOCH3

CAS RN 146195-69-9

IUPAC Name: (3,3-diethoxyprop-1-yn-1-yl)cyclopropane
Synonyms: (3,3-diethoxyprop-1-yn-1-yl)cyclopropane
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: WOSJTIAXBXVXLF-UHFFFAOYSA-N
SMILES: CCOC(OCC)C#CC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results