accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

N(R)N(R)O(S)(R)NNOHNH2HOHOF

CAS RN 146-78-1

IUPAC Name: 2-(6-amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: fludarabine
Molecular Weight (g/mol): 285.24
Molecular Formula: C10H12FN5O4
InChi Key: HBUBKKRHXORPQB-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC(F)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)