accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14589-33-4

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHHo

CAS RN 14589-33-4

IUPAC Name: tris(4-hydroxypent-3-en-2-one) holmium
Synonyms: tris(4-hydroxypent-3-en-2-one) holmium
Molecular Weight (g/mol): 465.28
Molecular Formula: C15H24HoO6
InChi Key: UPWLAELMJOPYAE-UHFFFAOYSA-N
SMILES: [Ho].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results