accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14548-39-1

BrO

CAS RN 14548-39-1

IUPAC Name: 6-bromo-2,3-dihydro-1H-inden-1-one
Synonyms: 6-bromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: SEQHEDQNODAFIU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2CCC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results