accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 145100-51-2

IUPAC Name: N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Synonyms: comins' reagent
Molecular Weight (g/mol): 392.67
Molecular Formula: C7H3ClF6N2O4S2
InChi Key: TUFGVZMNGTYAQD-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)N(C1=CC=C(Cl)C=N1)S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)