accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14507-19-8

OHOHOHLa

CAS RN 14507-19-8

IUPAC Name: lanthanum(3+) trihydroxide
Synonyms: lanthanum hydroxide
Molecular Weight (g/mol): 189.93
Molecular Formula: H3LaO3
InChi Key: YXEUGTSPQFTXTR-UHFFFAOYSA-K
SMILES: [OH-].[OH-].[OH-].[La+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results