accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BSOONHH3CHOOH

CAS RN 1449142-47-5

IUPAC Name: {3-[(2-methylphenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[(2-methylphenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 291.13
Molecular Formula: C13H14BNO4S
InChi Key: UBCMWUGADBYWIT-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NS(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results