accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1449142-43-1

BOHNClClFHOOH

CAS RN 1449142-43-1

IUPAC Name: {5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 327.93
Molecular Formula: C13H9BCl2FNO3
InChi Key: QBXYLSUKPFQIGC-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results