accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FNH2F

CAS RN 144851-63-8

IUPAC Name: 2,6-difluoro-3-methylaniline
Synonyms: 2,6-difluoro-3-methylaniline
Molecular Weight (g/mol): 143.14
Molecular Formula: C7H7F2N
InChi Key: ZMNJSSZHDRBGNN-UHFFFAOYSA-N
SMILES: CC1=C(F)C(N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results