accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14483-18-2

NOOONOOONOOOH2OH2OH2OH2OH2OHo

CAS RN 14483-18-2

IUPAC Name: holmium(3+) pentahydrate trinitrate
Synonyms: holmium(3+) pentahydrate trinitrate
Molecular Weight (g/mol): 441.02
Molecular Formula: H10HoN3O14
InChi Key: BJUGWWDCFYEYOA-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Ho+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)