accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14474-33-0

ScIII

CAS RN 14474-33-0

IUPAC Name: scandium(3+) triiodide
Synonyms: scandium triiodide
Molecular Weight (g/mol): 425.67
Molecular Formula: I3Sc
InChi Key: HUIHCQPFSRNMNM-UHFFFAOYSA-K
SMILES: [Sc+3].[I-].[I-].[I-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results