accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 144734-36-1

NOFOOOHHO

CAS RN 144734-36-1

IUPAC Name: 4-(4-fluorobenzoyl)-1-(4-phenylbutyl)piperidine; oxalic acid
Synonyms: 4-(4-fluorobenzoyl)-1-(4-phenylbutyl)piperidine; oxalic acid
Molecular Weight (g/mol): 429.49
Molecular Formula: C24H28FNO5
InChi Key: VUJYJCRJPFMHEM-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.FC1=CC=C(C=C1)C(=O)C1CCN(CCCCC2=CC=CC=C2)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results