accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiH3CCH3NO

CAS RN 14468-90-7

IUPAC Name: 1-(trimethylsilyl)pyrrolidin-2-one
Synonyms: 1-(trimethylsilyl)pyrrolidin-2-one
Molecular Weight (g/mol): 157.29
Molecular Formula: C7H15NOSi
InChi Key: LUBVCBITQHEVCJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1CCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)