accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 144625-51-4

H3CNOONHNSH3COO

CAS RN 144625-51-4

IUPAC Name: [1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 1-methyl-1H-indole-3-carboxylate
Synonyms: [1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 1-methylindole-3-carboxylate
Molecular Weight (g/mol): 393.5
Molecular Formula: C19H27N3O4S
InChi Key: MOZPSIXKYJUTKI-UHFFFAOYSA-N
SMILES: CN1C=C(C(=O)OCC2CCN(CCNS(C)(=O)=O)CC2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results