accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14456-51-0

BrBrBrTm

CAS RN 14456-51-0

IUPAC Name: thulium(3+) tribromide
Synonyms: thulium(3+) tribromide
Molecular Weight (g/mol): 408.65
Molecular Formula: Br3Tm
InChi Key: HQSWGSFQSCMHFQ-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Tm+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results