accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14456-47-4

BrBrBrTb

CAS RN 14456-47-4

IUPAC Name: terbium(3+) tribromide
Synonyms: terbium(III) bromide
Molecular Weight (g/mol): 398.64
Molecular Formula: Br3Tb
InChi Key: AZNZWHYYEIQIOC-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Tb+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results