accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHFFHCl

CAS RN 144230-52-4

IUPAC Name: 4,4-difluoropiperidine hydrochloride
Synonyms: 4,4-difluoropiperidine hydrochloride
Molecular Weight (g/mol): 157.59
Molecular Formula: C5H10ClF2N
InChi Key: OABUKBBBSMNNPM-UHFFFAOYSA-N
SMILES: Cl.FC1(F)CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)