accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 144036-17-9

IUPAC Name: 3-(cyclopentyloxy)-4-methoxybenzoic acid
Synonyms: 3-(cyclopentyloxy)-4-methoxybenzoic acid
Molecular Weight (g/mol): 236.27
Molecular Formula: C13H16O4
InChi Key: RVADCQWIQKYXBJ-UHFFFAOYSA-N
SMILES: COC1=C(OC2CCCC2)C=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results