accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOFNF

CAS RN 143879-77-0

IUPAC Name: 2,6-difluoro-3-nitrobenzonitrile
Synonyms: 2,6-difluoro-3-nitrobenzonitrile
Molecular Weight (g/mol): 184.1
Molecular Formula: C7H2F2N2O2
InChi Key: JQHDRFKDZHGEBQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(C#N)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results