accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FFFFOOHNH2

CAS RN 1435806-24-8

IUPAC Name: 2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 265.21
Molecular Formula: C11H11F4NO2
InChi Key: ZAEBGVLZTPXBJV-UHFFFAOYNA-N
SMILES: CC1=CC(CC(N)C(O)=O)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results