accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1435-49-0

FClCl

CAS RN 1435-49-0

IUPAC Name: 1,2-dichloro-4-fluorobenzene
Synonyms: 1,2-dichloro-4-fluorobenzene
Molecular Weight (g/mol): 164.99
Molecular Formula: C6H3Cl2F
InChi Key: QSDKXMVGRLVIQV-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results